Calcium Sensor Dye eFluor® 514 , eBioscience
65-0859-39 65-0859-70
102056-984EA 706.15 USD
102056-984 102056-986
Calcium Sensor Dye eFluor® 514 , eBioscience
Protein Linking and Labeling Systems
eFluor™ 514 Calcium Sensor Dye is a membrane-permeable dye used for monitoring changes in intracellular free calcium concentrations in the cell using fluorescence microscopy, flow cytometry, fluorescence spectroscopy and fluorescence microplate readers. Once eFluor™ 514 Calcium Sensor Dye enters the cell, cellular esterases cleave the AM group yielding a membrane-impermeable dye fluorescing at ~520 nm . eFluor™ 514 Calcium Sensor Dye, like Fluo-3 and Fluo-4, is a commonly used dye among the visible light-excitable calcium indicators but with increased cellular uptake (even at room temperature) and brightness. calcium binding affinity: Kd = 232 nM Molecular Weight: approximately 1100Peak Excitation: 490 nmPeak Emission: 514 nm eFluor™ 514 Calcium Sensor Dye should be reconstituted in high-quality, freshly opened DMSO. Recommended concentration: 2-5 mM.Once reconstituted, it should be protected from light and stored dessicated at -20°C; avoid freeze-thawing.