Anti-GARP Rat Antibody [clone: YGIC86]

Supplier: Affymetrix, Inc.
46-9891-80 46-9891-82
102054-552EA 176 USD
102054-552 102054-554
Anti-GARP Rat Antibody [clone: YGIC86]
Antibodies
The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predominant (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta. When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells. YGIC86 does not cross-react to rat GARP. Applications Reported: This YGIC86 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YGIC86 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4 °C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.

Flow Cytometry: 0.25 µg/test

Precaución: Regulatory status: RUO

Type: Primary
Antigen: GARP
Clonality: Monoclonal
Clone: YGIC86
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR