Anti-CD134 (OX40) Mouse Monoclonal Antibody [clone: ACT35 (ACT-35)]

Supplier: Affymetrix, Inc.
17-1347-41 17-1347-42
102051-274EA 137.85 USD
102051-274 102051-276
Anti-CD134 (OX40) Mouse Monoclonal Antibody [clone: ACT35 (ACT-35)]
Antibodies
The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50 kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4^+ T cell compartment. Applications Reported: The ACT35 (ACT-35) antibody has been reported for use in flow cytometric analysis. Applications Tested: This ACT35 (ACT-35) antibody has been pre-titrated and tested by flow cytometric analysis of 3-day PHA-stimulated normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that this receptor promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation.

Flow Cytometry: 5 µL (0.125 µg)/test

Precaución: Regulatory status: RUO

Type: Primary
Antigen: TNFRSF4
Clonality: Monoclonal
Clone: ACT35 (ACT-35)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR