Cell Stimulation Cocktail (500X) , eBioscience
00-4970-93 00-4970-03
102038-844EA 181.54 USD
102038-844 102038-842
Cell Stimulation Cocktail (500X) , eBioscience
Assays Cellular Assays
The Cell Stimulation Cocktail (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin
Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. This cocktail can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays. It can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression.