Glutaraldehyde 25% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
|
Danger
|
Synonyms:
Glutaral, Glutardialdehyde, 1,5-Pentanedial
Management of Change (MOC) category = N
2127-01
2127-03
2127-07
JT2127-1CS
3374.25
USD
JT2127-1
JT2127-3
JT2127-07
Glutaraldehyde 25% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Glutaraldehyde
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 101 °C (1013 hPa) Melting Pt: -6 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: ≥ 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007025 CAS Number: 111-30-8 EINECS: 203-856-5 UN: 3265 ADR: 6.1,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Assay (C₅H₈O₂) (UV Analysis) | 23 - 31 % |
pH at 25°C | 3.1 - 4.5 |
Clarity of 1% Solution | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...