Hydroxyethyl cellulose for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Hydroxyethyl Cellulose
M-Clarity™ quality level = MQ200
8.22068.0500
8.22068.0100
EM822068.0500EA
151
USD
EM822068.0500
EM8.22068.0100
Hydroxyethyl cellulose for synthesis, Sigma-Aldrich®
Hydroxyethyl cellulose
Storage Temperature:
Ambient |
MDL Number:
MFCD00072770 CAS Number: 9004-62-0 |
Specification Test Results
Assay (cellulose ether) | ≥92% |
Bulk Density | 350 kg/m3 |
pH | (10 g/l, H₂O, 20 °C) neutral |
Ignition Point | >360 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...