Corning® Polysucrose 400, Corning
Supplier: Corning
61-196-RM 61-196-RO
45000-730EA 285.27 USD
45000-730 45000-732
Corning® Polysucrose 400, Corning
Media for Microbiology Dehydrated Media for Microbiology
Polysucrose 400 is a synthetic polymer of sucrose and epichlorohydrin used for the density gradient separation of cells, cell membranes and virus cells
It can also be used to isolate different organelles. This nonionic compound is provided in powder form.