Ace-Thred Connectors, Stainless Steel, Ace Glass Incorporated
Supplier: Ace Glass
7644-70 7644-72 7644-76 7644-79 7644-81
89059-960EA 166.15 USD
89059-960 89059-962 89059-964 89059-966 89059-968
Ace-Thred Connectors, Stainless Steel, Ace Glass Incorporated
Fittings and Connectors
These internally threaded connectors can be used with couplings and plugs for positive pressure o-ring seal
Fabricated from 18-8 type 303 free machining stainless steel. O.D value represents the value of "Thread O.D".
Información para pedidos: Fittings and bushings for all sizes must be ordered separately.