LumiGLO Reserve® Chemiluminescent Substrate, SeraCare
Supplier: KPL
LumiGLO Reserve is a luminol-based chemiluminescent substrate designed for rapid and sensitive detection of peroxidase-labeled conjugates
LumiGLO Reserve has a dynamic range that is linear for a longer period, allowing for multiple exposures. An effective wash solution is included with the substrate.
It provides maximum sensitivity in blotting applications by allowing detection in the femtogram range. The substrate is ideal for detecting protein expressed in low abundance. Strong signal is achieved with the use of reduced amounts of precious target and antibodies.