Michel-Miller Chromatography Column, Ace Glass Incorporated
Supplier: Ace Glass
5820-57 5820-06 5820-18 5820-22 5820-36 5820-39
89051-958EA 270.74 USD
89051-958 89052-336 89052-364 89052-368 89052-376 89052-380
Michel-Miller Chromatography Column, Ace Glass Incorporated
Chromatography Columns
This column is designed for high performance, low pressure liquid chromatography (HP/LPLC) when used with appropriate PTFE fittings, and it is epoxy coated externally for added protection against scratching.
  • Fabricated from borosilicate glass
It is chemically resistant to dilute acids, most salts, and solvents. The maximum temperature limit is 235 °C. Matching Ace-Thred joints at each end. Column has straight-thru ends for use a packing column or operation column. Note the thread size when ordering so you can also order the proper size fittings. Column is intended for operation at elevated pressures and should always be used with plastic safety shields.
Precaución: Columns are for chromatography use only. Not intended for use with 40 micron or smaller packing, which will cause operating pressures to exceed safe limits. ALWAYS USE A SAFETY SHIELD. For custom columns, contact your Avantor representative.