gMAX Genomic DNA Kit, IBI Scientific
Supplier: IBI Scientific
IB47280 IB47281 IB47282
97060-568PK 32.46 USD
97060-568 97060-570 97060-572
gMAX Genomic DNA Kit, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
IBI's gMax MINI Genomic DNA Extraction/Purification Kit provides a fast and efficient method for purifying total DNA (including genomic, mitochondrial and viral DNA) from whole blood (fresh/frozen), tissue, buccal swab, and amniotic fluid
Note: For research use only.
The gMax MINI Kit allows the user to incubate tissue samples over-night, to make better use of valuable lab time. The gMax kit also includes Proteinase K powder that can be mixed with DI water and used during the cell lysing step for both tissue and blood samples. Chaotropic salt is used to lyse cells and degrade protein, allowing DNA to bind to the glass fiber matrix of the spin column. Contaminants are removed using a wash buffer (containing ethanol), and the purified genomic DNA is eluted by a low salt elution buffer or TE. The entire procedure can be completed in less than 30 minutes (after lysing) without phenol/chloroform extraction or alcohol precipitation, with an average DNA yield of 6µg from 200µl of of whole human blood. Purified DNA, with approximately 20-30Kb is suitable for use in PCR or other enzymatic reactions. The sample size for the gMax Kit is 50-200µl of whole blood (fresh or frozen), up to 25mg of tissue, up to 25mg of parafin-embedded tissue or buccal swab, and up to 15ml of amniotic fluid.
Precaución: Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.