4-(Trifluoromethyl)pyridine-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)picolinaldehyde

070807-500MG 070807-1G
101946-404EA 707.69 USD
101946-404 101946-406
4-(Trifluoromethyl)pyridine-2-carbaldehyde ≥95%
4-(Trifluoromethyl)pyridine-2-carbaldehyde

Hazard: Irritant/Refrigerate

Formula: C₇H₄F₃NO
MDL Number: MFCD11848102
CAS Number: 132470-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR