2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol ≥95%

Supplier: Matrix Scientific

070780-500MG 070780-1G
101946-300EA 403.08 USD
101946-300 101946-302
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol ≥95%
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazolidin-4-ol

Hazard: Irritant

Formula: C₉H₈ClF₃N₂O₂
MDL Number: MFCD21333205
CAS Number: 1357147-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR