3',5'-Dichloroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3,5-Dichlorophenyl)ethanone

070761-1G 070761-5G
101946-242EA 23.08 USD
101946-242 101946-244
3',5'-Dichloroacetophenone ≥98%
3',5'-Dichloroacetophenone

Hazard: Irritant

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
MDL Number: MFCD00045189
CAS Number: 14401-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR