1-(Triisopropylsilyl)pyrrole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Triisopropylsilyl)-1H-pyrrole

070759-1G 070759-5G 070759-25G
101946-236EA 38.46 USD
101946-236 101946-238 101946-240
1-(Triisopropylsilyl)pyrrole ≥97%
1-(Triisopropylsilyl)pyrrole

Hazard: Irritant/Moisture Sensitive

Formula: C₁₃H₂₅NSi
MW: 223.44 g/mol
Boiling Pt: 78 °C (0.4 mmHg)
Density: 0.904
Flash Pt: 106 °C
MDL Number: MFCD00054932
CAS Number: 87630-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR