5-Oxo-2-azaspiro[3.3]heptane-2-carboxylicacid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-oxo-2-azaspiro[3.3]heptane-2-carboxylate

070752-100MG 070752-250MG 070752-500MG
101946-208EA 930.77 USD
101946-208 101946-210 101946-212
5-Oxo-2-azaspiro[3.3]heptane-2-carboxylicacid tert-butyl ester ≥95%
5-Oxo-2-azaspiro[3.3]heptane-2-carboxylicacid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MW: 211.26 g/mol
MDL Number: MFCD17016196
CAS Number: 1251020-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR