3-(Nitromethyl)oxetane ≥95%

Supplier: Matrix Scientific

070751-100MG 070751-250MG 070751-500MG
101946-202EA 1396.92 USD
101946-202 101946-204 101946-206
3-(Nitromethyl)oxetane ≥95%
3-(Nitromethyl)oxetane

Hazard: Irritant

Formula: C₄H₇NO₃
MDL Number: MFCD12547196
CAS Number: 1313739-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR