1-Oxa-5-azaspiro[2.4]heptane-5-carboxylic acid tert-butyl ester ≥96%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-oxa-5-azaspiro[2.4]heptane-5-carboxylate

070749-250MG 070749-1G
101946-192EA 584.54 USD
101946-192 102551-772
1-Oxa-5-azaspiro[2.4]heptane-5-carboxylic acid tert-butyl ester ≥96%
1-Oxa-5-azaspiro[2.4]heptane-5-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD07779387
CAS Number: 301226-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR