2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid ≥96%

Supplier: Matrix Scientific

070747-100MG 070747-250MG 070747-500MG
101946-180EA 1396.92 USD
101946-180 101946-182 101946-184
2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid ≥96%
2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD18633612
CAS Number: 1009120-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR