1,7-Diazaspiro[3.5]nonane-1-carboxylic acid tert-butyl ester ≥94%

Supplier: Matrix Scientific

070746-250MG 070746-500MG
101946-176EA 830.77 USD
101946-176 101946-178
1,7-Diazaspiro[3.5]nonane-1-carboxylic acid tert-butyl ester ≥94%
1,7-Diazaspiro[3.5]nonane-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD14581196
CAS Number: 1216936-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR