3-(Bromomethyl)-3-fluorooxetane ≥95%

Supplier: Matrix Scientific

070745-100MG 070745-250MG 070745-500MG
101946-168EA 584.62 USD
101946-168 101946-170 101946-172
3-(Bromomethyl)-3-fluorooxetane ≥95%
3-(Bromomethyl)-3-fluorooxetane

Hazard: Irritant

Formula: C₄H₆BrFO
MDL Number: MFCD18909285
CAS Number: 865451-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR