Ethyl-2-(1-butylimidazol-2-yl)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

070741-1G 070741-5G
101946-148EA 640 USD
101946-148 101946-150
Ethyl-2-(1-butylimidazol-2-yl)-2-oxoacetate ≥95%
Ethyl-2-(1-butylimidazol-2-yl)-2-oxoacetate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MDL Number: MFCD19443898
CAS Number: 1313739-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR