Ethyl-2-(1-methylbenzimidazol-2-yl)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

070739-1G 070739-5G
101946-140EA 815.38 USD
101946-140 101946-142
Ethyl-2-(1-methylbenzimidazol-2-yl)-2-oxoacetate ≥95%
Ethyl-2-(1-methylbenzimidazol-2-yl)-2-oxoacetate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₃
MDL Number: MFCD13195937
CAS Number: 66155-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR