2-Chloro-1-cyclobutylbutane-1,3-dione ≥95%

Supplier: Matrix Scientific

070738-250MG 070738-500MG 070738-1G
101946-134EA 1338.46 USD
101946-134 101946-136 101946-138
2-Chloro-1-cyclobutylbutane-1,3-dione ≥95%
2-Chloro-1-cyclobutylbutane-1,3-dione

Hazard: Irritant

Formula: C₈H₁₁ClO₂
MDL Number: MFCD19443894
CAS Number: 1020732-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR