1-Cyclobutylbutane-1,3-dione ≥95%

Supplier: Matrix Scientific

070737-500MG 070737-1G 070737-5G
101946-128EA 815.38 USD
101946-128 101946-130 101946-132
1-Cyclobutylbutane-1,3-dione ≥95%
1-Cyclobutylbutane-1,3-dione

Hazard: Irritant

Formula: C₈H₁₂O₂
MDL Number: MFCD11553136
CAS Number: 1020732-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR