2-Chloro-1-cyclopropyl-1,3-butanedione ≥95%

Supplier: Matrix Scientific

070736-500MG 070736-1G 070736-5G
101946-122EA 349.23 USD
101946-122 101946-124 101946-126
2-Chloro-1-cyclopropyl-1,3-butanedione ≥95%
2-Chloro-1-cyclopropyl-1,3-butanedione

Hazard: Irritant

Formula: C₇H₉ClO₂
MDL Number: MFCD16875450
CAS Number: 473924-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR