2-Chloro-1,3-dicyclopropylpropane-1,3-dione ≥96%
Supplier: Matrix Scientific
070735-500MG
070735-1G
070735-5G
101946-116EA
640
USD
101946-116
101946-118
101946-120
2-Chloro-1,3-dicyclopropylpropane-1,3-dione ≥96%
2-Chloro-1,3-dicyclopropylpropane-1,3-dione
Hazard: Irritant
Formula:
C₉H₁₁ClO₂ |
MDL Number:
MFCD13172692 CAS Number: 473924-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...