1,3-Dicyclopropylpropane-1,3-dione ≥96%

Supplier: Matrix Scientific

070734-500MG 070734-1G 070734-5G
101946-110EA 349.23 USD
101946-110 101946-112 101946-114
1,3-Dicyclopropylpropane-1,3-dione ≥96%
1,3-Dicyclopropylpropane-1,3-dione

Hazard: Irritant

Formula: C₉H₁₂O₂
MDL Number: MFCD18330671
CAS Number: 30923-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR