endo-3-(Boc-amino)-8-azabicyclo[3.2.1]octane ≥95%

Supplier: Matrix Scientific

070733-100MG 070733-250MG 070733-500MG
101946-104EA 180 USD
101946-104 101946-106 101946-108
endo-3-(Boc-amino)-8-azabicyclo[3.2.1]octane ≥95%
endo-3-(Boc-amino)-8-azabicyclo[3.2.1]octane

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD16660224
CAS Number: 132234-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR