5,7-Diiodo-2,3-dihydrothieno[3,4-b][1,4]dioxine ≥95%

Supplier: Matrix Scientific

070731-250MG 070731-500MG 070731-1G
101946-092EA 524.62 USD
101946-092 101946-094 101946-096
5,7-Diiodo-2,3-dihydrothieno[3,4-b][1,4]dioxine ≥95%
5,7-Diiodo-2,3-dihydrothieno[3,4-b][1,4]dioxine

Hazard: Irritant

Formula: C₆H₄I₂O₂S
MW: 393.97 g/mol
MDL Number: MFCD19443896
CAS Number: 640737-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR