1,1-Bis(iodomethyl)cyclobutane ≥95%

Supplier: Matrix Scientific

070729-500MG 070729-1G
101946-082EA 1221.54 USD
101946-082 101946-084
1,1-Bis(iodomethyl)cyclobutane ≥95%
1,1-Bis(iodomethyl)cyclobutane

Hazard: Irritant

Formula: C₆H₁₀I₂
MDL Number: MFCD18839256
CAS Number: 65478-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR