5-Bromo-2-(trimethylsilyl)pyridine ≥95%

Supplier: Matrix Scientific

070726-1G 070726-5G 070726-25G
101946-070EA 1192.31 USD
101946-070 101946-072 101946-074
5-Bromo-2-(trimethylsilyl)pyridine ≥95%
5-Bromo-2-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₈H₁₂BrNSi
MDL Number: MFCD18839255
CAS Number: 291312-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR