1,4-Thiazepane-1,1-dioxidetrifluoroacetate ≥95%

Supplier: Matrix Scientific

070724-250MG 070724-500MG
101946-062EA 1978.46 USD
101946-062 101946-064
1,4-Thiazepane-1,1-dioxidetrifluoroacetate ≥95%
1,4-Thiazepane-1,1-dioxidetrifluoroacetate

Hazard: Irritant

Formula: C₈H₁₄F₃NO₃S
MW: 261.26 g/mol
MDL Number: MFCD19443977
CAS Number: 756815-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR