3-(Methoxymethylene)oxetane ≥97%

Supplier: Matrix Scientific

070718-250MG 070718-500MG 070718-1G
101946-038EA 1106.15 USD
101946-038 101946-040 101946-042
3-(Methoxymethylene)oxetane ≥97%
3-(Methoxymethylene)oxetane

Hazard: Irritant

Formula: C₅H₈O₂
MDL Number: MFCD18839252
CAS Number: 1313739-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR