S-(2-Chloroallyl)thioacetate ≥90%
Supplier: Matrix Scientific
Synonyms:
Ethanethioic acid s-(2-chloro-2-propen-1-yl)ester
070717-1G
070717-5G
101946-034EA
233.85
USD
101946-034
101946-036
S-(2-Chloroallyl)thioacetate ≥90%
S-(2-Chloroallyl)thioacetate
Hazard: Irritant
Formula:
C₅H₇ClOS MW: 150.63 g/mol |
MDL Number:
MFCD18839251 CAS Number: 24891-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...