tert-Butyl-3-tert-butylsulfinyliminoazetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

070716-250MG 070716-500MG 070716-1G
101946-028EA 930.77 USD
101946-028 101946-030 101946-032
tert-Butyl-3-tert-butylsulfinyliminoazetidine-1-carboxylate ≥95%
tert-Butyl-3-tert-butylsulfinyliminoazetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₃S
MDL Number: MFCD18839250
CAS Number: 1291487-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR