4,7-Diaza-1H-indazole ≥97%

Supplier: Matrix Scientific

070715-500MG 070715-1G 070715-5G
101946-022EA 524.62 USD
101946-022 101946-024 101946-026
4,7-Diaza-1H-indazole ≥97%
4,7-Diaza-1H-indazole

Hazard: Irritant

Formula: C₅H₄N₄
MW: 120.1121 g/mol
MDL Number: MFCD09999207
CAS Number: 272-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR