3,3-Oxetanedimethanol ≥95%

Supplier: Matrix Scientific

070707-250MG 070707-500MG 070707-1G
101945-992EA 396.23 USD
101945-992 101945-994 101945-996
3,3-Oxetanedimethanol ≥95%
3,3-Oxetanedimethanol

Hazard: Irritant

Formula: C₅H₁₀O₃
MDL Number: MFCD00463708
CAS Number: 2754-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR