Methyl-5-(methoxymethyl)-3-methylisoxazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

070706-500MG 070706-1G
101945-988EA 930.77 USD
101945-988 101945-990
Methyl-5-(methoxymethyl)-3-methylisoxazole-4-carboxylate ≥95%
Methyl-5-(methoxymethyl)-3-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₄
MDL Number: MFCD18839246
CAS Number: 1108712-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR