5-(Methoxymethyl)-3-methylisoxazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

070705-500MG 070705-1G
101945-984EA 1221.54 USD
101945-984 101945-986
5-(Methoxymethyl)-3-methylisoxazole-4-carboxylic acid ≥95%
5-(Methoxymethyl)-3-methylisoxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₉NO₄
MDL Number: MFCD18839245
CAS Number: 1108712-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR