1-Benzyl-4-bromo-3,6-dihydro-2H-pyridine ≥95%

Supplier: Matrix Scientific

070700-250MG 070700-500MG 070700-1G
101945-968EA 466.15 USD
101945-968 101945-970 101945-972
1-Benzyl-4-bromo-3,6-dihydro-2H-pyridine ≥95%
1-Benzyl-4-bromo-3,6-dihydro-2H-pyridine

Hazard: Irritant

Formula: C₁₂H₁₄BrN
MDL Number: MFCD11501647
CAS Number: 175347-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR