N-(1-Benzhydrylazetidin-3-ylidene)-2-methylpropane-2-sulfinamide ≥97%

Supplier: Matrix Scientific

070699-250MG 070699-500MG 070699-1G
101945-962EA 930.77 USD
101945-962 101945-964 101945-966
N-(1-Benzhydrylazetidin-3-ylidene)-2-methylpropane-2-sulfinamide ≥97%
N-(1-Benzhydrylazetidin-3-ylidene)-2-methylpropane-2-sulfinamide

Hazard: Irritant

Formula: C₂₀H₂₄N₂OS
MW: 340.49 g/mol
MDL Number: MFCD18839242
CAS Number: 1263296-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR