1-Benzhydryl-3-(trifluoromethyl)azetidin-3-ol ≥96%

Supplier: Matrix Scientific

070696-100MG 070696-250MG 070696-500MG
101945-944EA 1221.54 USD
101945-944 101945-946 101945-948
1-Benzhydryl-3-(trifluoromethyl)azetidin-3-ol ≥96%
1-Benzhydryl-3-(trifluoromethyl)azetidin-3-ol

Hazard: Irritant

Formula: C₁₇H₁₆F₃NO
MDL Number: MFCD18839240
CAS Number: 848192-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR