Ethyl-2-(1-benzhydrylazetidin-3-ylidene) acetate ≥96%

Supplier: Matrix Scientific

070695-500MG 070695-1G
101945-940EA 815.38 USD
101945-940 101945-942
Ethyl-2-(1-benzhydrylazetidin-3-ylidene) acetate ≥96%
Ethyl-2-(1-benzhydrylazetidin-3-ylidene) acetate

Hazard: Irritant

Formula: C₂₀H₂₁NO₂
MDL Number: MFCD11227131
CAS Number: 158602-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR