4-Chloro-6-methyl-2-(methylthio)pyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-chloro-6-methyl-2-(methylsulphanyl)pyrimidine, 4-Chloro-6-methyl-2-methylthiopyrimidine

070693-1G 070693-5G
101945-936EA 41.54 USD
101945-936 101945-938
4-Chloro-6-methyl-2-(methylthio)pyrimidine ≥98%
4-Chloro-6-methyl-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₆H₇ClN₂S
MW: 174.65 g/mol
MDL Number: MFCD00023233
CAS Number: 17119-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR