1-Oxa-8-azaspiro[4.5]decane hydrochloride ≥94%

Supplier: Matrix Scientific

070692-250MG 070692-500MG 070692-1G
101945-930EA 570.77 USD
101945-930 101945-932 101945-934
1-Oxa-8-azaspiro[4.5]decane hydrochloride ≥94%
1-Oxa-8-azaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO
MDL Number: MFCD11042294
CAS Number: 3970-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR