(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: [3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl]methanol, [3-(Bromomethyl)-1-tosylazetidin-3-yl]methanol

070686-500MG 070686-1G
101945-900EA 1106.15 USD
101945-900 101945-902
(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol ≥95%
(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₃S
MDL Number: MFCD18782897
CAS Number: 1041026-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR