3,3-Bis(bromomethyl)-1-(p-toluenesulfonyl)azetidine ≥90%

Supplier: Matrix Scientific

070685-500MG 070685-1G
101945-896EA 1396.92 USD
101945-896 101945-898
3,3-Bis(bromomethyl)-1-(p-toluenesulfonyl)azetidine ≥90%
3,3-Bis(bromomethyl)-1-(p-toluenesulfonyl)azetidine

Hazard: Irritant

Formula: C₁₂H₁₅Br₂NO₂S
MW: 397.14 g/mol
MDL Number: MFCD18782896
CAS Number: 1041026-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR