1-Benzyl-1,6-diazaspiro[3.3]heptane hemioxalate ≥95%
Supplier: Matrix Scientific
070683-100MG
070683-250MG
070683-500MG
101945-886EA
1047.69
USD
101945-886
101945-888
101945-890
1-Benzyl-1,6-diazaspiro[3.3]heptane hemioxalate ≥95%
1-Benzyl-1,6-diazaspiro[3.3]heptane hemioxalate
Hazard: Irritant
Formula:
C₂₆H₃₄N₄O₄ |
MDL Number:
MFCD18782895 CAS Number: 1223573-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...