6-(p-Toluenesulfonyl)-2-oxa-6-azaspiro[3.3]heptane ≥94%

Supplier: Matrix Scientific

Synonyms: 6-Tosyl-2-oxa-6-azaspiro[3.3]heptane

070680-1G 070680-5G
101945-876EA 640 USD
101945-876 101945-878
6-(p-Toluenesulfonyl)-2-oxa-6-azaspiro[3.3]heptane ≥94%
6-(p-Toluenesulfonyl)-2-oxa-6-azaspiro[3.3]heptane

Hazard: Irritant

Formula: C₁₂H₁₅NO₃S
MW: 253.32 g/mol
MDL Number: MFCD13180735
CAS Number: 13573-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR